MilliporeSigma
  • Transformation of LTP gene into Brassica napus to enhance its resistance to Sclerotinia sclerotiorum.
Transformation of LTP gene into Brassica napus to enhance its resistance to Sclerotinia sclerotiorum.
Genetika (2013-07-23)
Y Fan, K Du, Y Gao, Y Kong, C Chu, V Sokolov, Y Wang
ABSTRACT
Rapeseed (Brassica napus L.) is one of the most important economic crops worldwide, and Sclerotinia sclerotiorum is the most dangerous disease that affects its yield greatly. Lipid transfer protein (LTP) has broad-spectrum anti-bacterial and fungal activities. In this study, B. napus was transformed using Agrobacterium tumefaciens harboring the plasmid-containing LTP gene to study its possible capability of increasing plant's resistance. First, we optimized the petiole genetic transformation system by adjusting the days of explants, bacterial concentrations, ratio of hormones, and cultivating condition. Second, we obtained 8 positive plants by PCR analysis of T0 generation. The PCR results of T1 generation were positive, indicating that the LTP gene had been integrated into B. napus. Third, T1 transgenic plants inoculated by detached leaves with mycelia of S. sclerotiorum showed better disease resistance than non-transformants. Oxalic acid belongs to secondary metabolites of S. sclerotiorum, and several studies have demonstrated that the resistance of rapeseed to oxalic acid is significantly consistent with its resistance to S. sclerotiorum. The result from the seed germination assay showed that when T1 seeds were exposed to oxalic acid stress, their germination rate was evidently higher than that of non-transformant seeds. In addition, we measured some physiological changes in T1 plants and control plants under oxalic acid stress. The results showed that T1 transgenic plants had lower malondialdehyde (MDA) content, higher super oxide dismutase (SOD), and peroxidase (POD) activities than non-transformants, whereas disease resistance was related to low MDA content and high SOD and POD activities.
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%