MilliporeSigma
  • Iron and citrate export by a major facilitator superfamily pump regulates metabolism and stress resistance in Salmonella Typhimurium.
Iron and citrate export by a major facilitator superfamily pump regulates metabolism and stress resistance in Salmonella Typhimurium.
Proceedings of the National Academy of Sciences of the United States of America (2013-07-04)
Elaine R Frawley, Marie-Laure V Crouch, Lacey K Bingham-Ramos, Hannah F Robbins, Wenliang Wang, Gerard D Wright, Ferric C Fang
ABSTRACT
The efficacy of antibiotics and host defenses has been linked to the metabolic and redox states of bacteria. In this study we report that a stress-induced export pump belonging to the major facilitator superfamily effluxes citrate and iron from the enteric pathogen Salmonella Typhimurium to arrest growth and ameliorate the effects of antibiotics, hydrogen peroxide, and nitric oxide. The transporter, formerly known as MdtD, is now designated IceT (iron citrate efflux transporter). Iron efflux via an iron-chelating tricarboxylic acid cycle intermediate provides a direct link between aerobic metabolism and bacterial stress responses, representing a unique mechanism of resistance to host defenses and antimicrobial agents of diverse classes.
Sigma-Aldrich
Streptonigrin from Streptomyces flocculus, ≥98%