MilliporeSigma
  • What a difference a hydroxyl makes: mutant IDH, (R)-2-hydroxyglutarate, and cancer.
What a difference a hydroxyl makes: mutant IDH, (R)-2-hydroxyglutarate, and cancer.
Genes & development (2013-05-01)
Julie-Aurore Losman, William G Kaelin
ABSTRACT
Mutations in metabolic enzymes, including isocitrate dehydrogenase 1 (IDH1) and IDH2, in cancer strongly implicate altered metabolism in tumorigenesis. IDH1 and IDH2 catalyze the interconversion of isocitrate and 2-oxoglutarate (2OG). 2OG is a TCA cycle intermediate and an essential cofactor for many enzymes, including JmjC domain-containing histone demethylases, TET 5-methylcytosine hydroxylases, and EglN prolyl-4-hydroxylases. Cancer-associated IDH mutations alter the enzymes such that they reduce 2OG to the structurally similar metabolite (R)-2-hydroxyglutarate [(R)-2HG]. Here we review what is known about the molecular mechanisms of transformation by mutant IDH and discuss their implications for the development of targeted therapies to treat IDH mutant malignancies.
Sigma-Aldrich
DL-α-Hydroxyglutaric acid disodium salt, ≥95.0% (GC)
Sigma-Aldrich
D-2-Hydroxyglutarate (D2HG) Assay Kit, sufficient for 200 fluorometric reactions