MilliporeSigma
  • Comparison of uncoupling activities of chlorophenoxy herbicides in rat liver mitochondria.
Comparison of uncoupling activities of chlorophenoxy herbicides in rat liver mitochondria.
Toxicology letters (1990-06-01)
L Zychlinski, S Zolnierowicz
ABSTRACT
The effects of the herbicides 2,4-dichlorophenoxyacetic acid (2,4-D), 2,4,5-trichlorophenoxyacetic acid (2,4,5-T), 4-chloro-2-methylphenoxyacetic acid (MCPA) and 2-(2,4,5-trichlorophenoxy)propionic acid (2,4,5-TP) on respiration and oxidative phosphorylation in rat liver mitochondria were examined in vitro. Respiration rates of glutamate, malate and succinate were investigated in the presence of each herbicide (0.1-4.0 mM). At lower concentrations, all herbicides stimulated state 4 respiration, decreased the respiratory control ratio and the ADP/O ratio. The respiration rate in state 3 and uncoupled state was unaffected. At higher concentrations all bioenergetic parameters, respiration in state 4, 3 and uncoupled state, as well as respiratory control ratio and ADP/O, were inhibited in a concentration-dependent manner. These data indicate that these herbicides alter energy metabolism in rat liver mitochondria by uncoupling of oxidative phosphorylation. 2,4,5-TP possesses the strongest uncoupling properties followed by 2,4,5-T, MCPA and 2,4-D in that order.
Product Description
Supelco
2-(2,4,5-Trichlorophenoxy)propionic acid, PESTANAL®, analytical standard