MilliporeSigma
  • ZnO nanorod optical disk photocatalytic reactor for photodegradation of methyl orange.
ZnO nanorod optical disk photocatalytic reactor for photodegradation of methyl orange.
Optics express (2013-04-03)
Yu Lim Chen, Li-Chung Kuo, Min Lun Tseng, Hao Ming Chen, Chih-Kai Chen, Hung Ji Huang, Ru-Shi Liu, Din Ping Tsai
ABSTRACT
A low-cost and efficient photocatalytic reactor for environmental treatment and green technology was presented. ZnO nanorods firmly growing on polycarbonate optical disk substrate are generally perpendicular to the substrate as the immobilized photocatalyst of the spinning disk reactor. The photocatalytic efficiency and durability of the ZnO nanorods are effectively demonstrated.
Sigma-Aldrich
Methyl Orange, for microscopy (Hist.), indicator (pH 3.0-4.4)
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %