MilliporeSigma
  • Plasticization effect of triacetin on structure and properties of starch ester film.
Plasticization effect of triacetin on structure and properties of starch ester film.
Carbohydrate polymers (2013-04-03)
Jie Zhu, Xiaoxi Li, Chen Huang, Ling Chen, Lin Li
ABSTRACT
The aim of this work was to evaluate the plasticizing effect of triacetin on the structure and properties of starch ester film and further establish the structure-property relationships. The presence of triacetin resulted in multiple structure changes of the film. The mobility of macromolecular chain was increased to form scattered crystallite during the film formation process. The amorphous region was enlarged to contain more triacetin squeezed from crystalline region. The plasticization of triacetin and restriction of crystallite oppositely influenced the mobility of macromolecular chains in different regions. The thermal stability of triacetin changed along with its fluctuant interaction with macromolecules. Comparatively, the enhanced ether bond and the restriction from crystalline regions on the mobility of the amorphous chain consequently improved the thermal stability of the film matrix. The interaction between triacetin and starch ester was essential to film forming but unexpectedly lowered the triacetin stability.
Sigma-Aldrich
Triacetin, 99%
Sigma-Aldrich
Triacetin, 99%, FCC, FG
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Glyceryl triacetate, ≥99%
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Glyceryl triacetate, ≥99.0% (GC)
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble