MilliporeSigma
  • Determination of dapoxetine in rat plasma by ultra-performance liquid chromatography-tandem mass spectrometry.
Determination of dapoxetine in rat plasma by ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-04-02)
Tae Kon Kim, In Sook Kim, Seok Hyun Hong, Yun Kyoung Choi, Hohyun Kim, Hye Hyun Yoo
ABSTRACT
In this study, we describe and validate a rapid and sensitive method for quantitation of dapoxetine in rat plasma by using ultra-performance liquid chromatography-electrospray ionization tandem mass spectrometry (UPLC-ESI/MS/MS). Plasma samples were prepared by protein precipitation with acetonitrile, and sildenafil was used as an internal standard (IS). The mobile phase consisted of 0.5% formic acid/acetonitrile (60:40, v/v); a C18 reversed-phase column (2.0 × 50 mm, 1.7 μm) was used for chromatographic separation. Multiple reaction monitoring (MRM) was used in the positive ion mode for mass spectrometric detection. The calibration curve for dapoxetine was linear (r(2)=0.999) in the concentration range of 1-500 ng/mL. The intra- and inter-day precision was between 3.8% and 8.3%, and the intra- and inter-day accuracy was between 101.1% and 109.0%. Dapoxetine was found to be stable in various conditions with the recoveries>87.0% (RSD <7.2%). The method was found to be specific, precise, and accurate, and no matrix effect was observed. Our results suggest that this method can be successfully applied in pharmacokinetic studies of dapoxetine in rat plasma.
Sigma-Aldrich
Dapoxetine hydrochloride, ≥98% (HPLC)