MilliporeSigma
  • Evaluation of glomerular filtration rate in cats with reduced renal mass and administered meloxicam and acetylsalicylic acid.
Evaluation of glomerular filtration rate in cats with reduced renal mass and administered meloxicam and acetylsalicylic acid.
American journal of veterinary research (2013-03-28)
Kathryn K Surdyk, Cathy A Brown, Scott A Brown
ABSTRACT
To determine whether administration of meloxicam or acetylsalicylic acid alters glomerular filtration rate (GFR) in cats with renal azotemia. 6 young adult cats. 3 sexually intact male cats and 3 sexually intact female cats had surgically reduced renal mass and azotemia comparable to International Renal Interest Society chronic kidney disease stages 2 and 3. Renal function was evaluated by measurement of serum creatinine concentration, urinary clearance of exogenously administered creatinine, and the urine protein-to-creatinine concentration ratio (UP:C). Measurements taken in cats receiving placebo at the beginning and end of the study were compared with results obtained at the end of 7 days of treatment with either meloxicam (0.2 mg/kg, SC, on day 1; 0.1 mg/kg, SC, on days 2 to 7) or acetylsalicylic acid (20 mg/kg, PO, on days 1, 4, and 7). No significant treatment effects on urinary clearance of exogenously administered creatinine, serum creatinine concentration, or UP:C were detected. Mean ± SEM serum creatinine concentration and urinary clearance of exogenously administered creatinine measurements following 7 days of treatment with meloxicam (serum creatinine concentration, 2.67 ± 0.17 mg/dL; urinary clearance of exogenously administered creatinine, 1.34 ± 0.08 mL/min/kg) and acetylsalicylic acid (serum creatinine concentration, 2.62 ± 0.12 mg/dL; urinary clearance of exogenously administered creatinine, 1.35 ± 0.07 mL/min/kg) were not significantly different from the mean baseline values for these variables (serum creatinine concentration, 2.77 ± 0.14 mg/dL; urinary clearance of exogenously administered creatinine, 1.36 ± 0.07 mL/min/kg). Neither meloxicam nor acetylsalicylic acid had a measurable effect on urinary clearance of exogenously administered creatinine, serum creatinine concentration, or UP:C. These results are consistent with the hypothesis that GFR of euvolemic cats with normal or reduced renal function is not dependent on cyclooxygenase function.
Product Description
Meloxicam, European Pharmacopoeia (EP) Reference Standard