MilliporeSigma
  • Fluorescent water-soluble perylenediimide-cored cationic dendrimers: synthesis, optical properties, and cell uptake.
Fluorescent water-soluble perylenediimide-cored cationic dendrimers: synthesis, optical properties, and cell uptake.
Chemical communications (Cambridge, England) (2013-03-27)
Zejun Xu, Bicheng He, Jie Shen, Wantai Yang, Meizhen Yin
ABSTRACT
Different generations of perylenediimide-cored dendrimers with peripheral amine groups were synthesized. All these water-soluble dendrimers could rapidly internalize into live cells with high efficacy of gene transfection and low cytotoxicity. Increasing dendrimer generation increased their ability for gene transfection.
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%