MilliporeSigma
  • Characterization of five ECF sigma factors in the genome of Pseudomonas syringae pv. syringae B728a.
Characterization of five ECF sigma factors in the genome of Pseudomonas syringae pv. syringae B728a.
PloS one (2013-03-22)
Poulami Basu Thakur, Vanessa L Vaughn-Diaz, Jessica W Greenwald, Dennis C Gross
ABSTRACT
Pseudomonas syringae pv. syringae B728a, a bacterial pathogen of bean, utilizes large surface populations and extracellular signaling to initiate a fundamental change from an epiphytic to a pathogenic lifestyle. Extracytoplasmic function (ECF) sigma (σ) factors serve as important regulatory factors in responding to various environmental signals. Bioinformatic analysis of the B728a genome revealed 10 ECF sigma factors. This study analyzed deletion mutants of five previously uncharacterized ECF sigma factor genes in B728a, including three FecI-type ECF sigma factors (ECF5, ECF6, and ECF7) and two ECF sigma factors placed in groups ECF11 and ECF18. Transcriptional profiling by qRT-PCR analysis of ECF sigma factor mutants was used to measure expression of their associated anti-sigma and outer membrane receptor proteins, and expression of genes associated with production of extracellular polysaccharides, fimbriae, glycine betaine and syringomycin. Notably, the B728aΔecf7 mutant displayed reduced swarming and had decreased expression of CupC fimbrial genes. Growth and pathogenicity assays, using a susceptible bean host, revealed that none of the tested sigma factor genes are required for in planta growth and lesion formation.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%