MilliporeSigma
  • Inhibition of mitochondrial aconitase by succination in fumarate hydratase deficiency.
Inhibition of mitochondrial aconitase by succination in fumarate hydratase deficiency.
Cell reports (2013-03-19)
Nicola Ternette, Ming Yang, Mahima Laroyia, Mitsuhiro Kitagawa, Linda O'Flaherty, Kathryn Wolhulter, Kaori Igarashi, Kaori Saito, Keiko Kato, Roman Fischer, Alexandre Berquand, Benedikt M Kessler, Terry Lappin, Norma Frizzell, Tomoyoshi Soga, Julie Adam, Patrick J Pollard
ABSTRACT
The gene encoding the Krebs cycle enzyme fumarate hydratase (FH) is mutated in hereditary leiomyomatosis and renal cell cancer (HLRCC). Loss of FH activity causes accumulation of intracellular fumarate, which can directly modify cysteine residues to form 2-succinocysteine through succination. We undertook a proteomic-based screen in cells and renal cysts from Fh1 (murine FH)-deficient mice and identified 94 protein succination targets. Notably, we identified the succination of three cysteine residues in mitochondrial Aconitase2 (ACO2) crucial for iron-sulfur cluster binding. We show that fumarate exerts a dose-dependent inhibition of ACO2 activity, which correlates with increased succination as determined by mass spectrometry, possibly by interfering with iron chelation. Importantly, we show that aconitase activity is impaired in FH-deficient cells. Our data provide evidence that succination, resulting from FH deficiency, targets and potentially alters the function of multiple proteins and may contribute to the dysregulated metabolism observed in HLRCC.
Sigma-Aldrich
Aconitase from porcine heart
Sigma-Aldrich
Fumarase from porcine heart, ammonium sulfate suspension, ≥300 units/mg protein (biuret)
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Fumaric acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland