MilliporeSigma
  • Adsorption of cesium from aqueous solution using agricultural residue--walnut shell: equilibrium, kinetic and thermodynamic modeling studies.
Adsorption of cesium from aqueous solution using agricultural residue--walnut shell: equilibrium, kinetic and thermodynamic modeling studies.
Water research (2013-03-14)
Dahu Ding, Yingxin Zhao, Shengjiong Yang, Wansheng Shi, Zhenya Zhang, Zhongfang Lei, Yingnan Yang
ABSTRACT
A novel biosorbent derived from agricultural residue - walnut shell (WS) is reported to remove cesium from aqueous solution. Nickel hexacyanoferrate (NiHCF) was incorporated into this biosorbent, serving as a high selectivity trap agent for cesium. Field emission scanning electron microscope (FE-SEM) and thermogravimetric and differential thermal analysis (TG-DTA) were utilized for the evaluation of the developed biosorbent. Determination of kinetic parameters for adsorption was carried out using pseudo first-order, pseudo second-order kinetic models and intra-particle diffusion models. Adsorption equilibrium was examined using Langmuir, Freundlich and Dubinin-Radushkevich adsorption isotherms. A satisfactory correlation coefficient and relatively low chi-square analysis parameter χ(2) between the experimental and predicted values of the Freundlich isotherm demonstrate that cesium adsorption by NiHCF-WS is a multilayer chemical adsorption. Thermodynamic studies were conducted under different reaction temperatures and results indicate that cesium adsorption by NiHCF-WS is an endothermic (ΔH° > 0) and spontaneous (ΔG° < 0) process.
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis