MilliporeSigma
  • Phase III randomized study of rituximab/carmustine, etoposide, cytarabine, and melphalan (BEAM) compared with iodine-131 tositumomab/BEAM with autologous hematopoietic cell transplantation for relapsed diffuse large B-cell lymphoma: results from the BMT CTN 0401 trial.
Phase III randomized study of rituximab/carmustine, etoposide, cytarabine, and melphalan (BEAM) compared with iodine-131 tositumomab/BEAM with autologous hematopoietic cell transplantation for relapsed diffuse large B-cell lymphoma: results from the BMT CTN 0401 trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-03-13)
Julie M Vose, Shelly Carter, Linda J Burns, Ernesto Ayala, Oliver W Press, Craig H Moskowitz, Edward A Stadtmauer, Shin Mineshi, Richard Ambinder, Timothy Fenske, Mary Horowitz, Richard Fisher, Marcie Tomblyn
ABSTRACT
This clinical trial evaluated standard-dose radioimmunotherapy with a chemotherapy-based transplantation regimen followed by autologous hematopoietic cell transplantation versus rituximab with the same regimen in patients with relapsed diffuse large B-cell lymphoma (DLBCL). Patients with chemotherapy-sensitive persistent or relapsed DLBCL were randomly assigned to receive iodine-131 tositumomab (dosimetric dose of 5 mCi on day -19 and therapeutic dose of 0.75 Gy on day -12), carmustine 300 mg/m(2) (day -6), etoposide 100 mg/m(2) twice daily (days -5 to -2), cytarabine 100 mg/m(2) twice daily (days -5 to -2), and melphalan 140 mg/m(2) (day -1; B-BEAM) or rituximab 375 mg/m(2) on days -19 and -12 and the same chemotherapy regimen (R-BEAM). Two hundred twenty-four patients were enrolled, with 113 patients randomly assigned to R-BEAM and 111 patients assigned to B-BEAM. Two-year progression-free survival (PFS) rates, the primary end point, were 48.6% (95% CI, 38.6% to 57.8%) for R-BEAM and 47.9% (95% CI, 38.2% to 57%; P = .94) for B-BEAM, and the 2-year overall survival (OS) rates were 65.6% (95% CI, 55.3% to 74.1%) for R-BEAM and 61% (95% CI, 50.9% to 69.9%; P = .38) for B-BEAM. The 100-day treatment-related mortality rates were 4.1% (95% CI, 0.2% to 8.0%) for R-BEAM and 4.9% (95% CI, 0.8% to 9.0%; P = .97) for B-BEAM. The maximum mucositis score was higher in the B-BEAM arm (0.72) compared with the R-BEAM arm (0.31; P < .001). The B-BEAM and R-BEAM regimens produced similar 2-year PFS and OS rates for patients with chemotherapy-sensitive relapsed DLBCL. No differences in toxicities other than mucositis were noted.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)