MilliporeSigma
  • Synthesis of indene frameworks via rhodium-catalyzed cascade cyclization of aromatic ketone and unsaturated carbonyl compounds.
Synthesis of indene frameworks via rhodium-catalyzed cascade cyclization of aromatic ketone and unsaturated carbonyl compounds.
Organic letters (2013-03-09)
Xian-Ying Shi, Chao-Jun Li
ABSTRACT
A novel rhodium(III)-catalyzed direct functionalization of the ortho-C-H bond of aromatic ketone derivatives and an intramolecular cyclization sequence produced indene derivatives in moderate to good yields. This cascade cyclization involves a conjugate addition of α,β-unsaturated ketone and subsequent aldol condensation. The reaction occurred efficiently in the presence of water and under an atmosphere of air.
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis