MilliporeSigma
  • Mutagenicity evaluation of HC Blue No. 1 and HC Blue No. 2, II. Effect on the in vitro induction of unscheduled DNA synthesis in rat, mouse, rabbit, hamster, and monkey primary hepatocytes.
Mutagenicity evaluation of HC Blue No. 1 and HC Blue No. 2, II. Effect on the in vitro induction of unscheduled DNA synthesis in rat, mouse, rabbit, hamster, and monkey primary hepatocytes.
Mutation research (1990-06-01)
L E Hill, J W Parton, G S Probst, M L Garriott
ABSTRACT
2 hair dyes, HC Blue No. 1 and HC Blue No. 2, were evaluated for the in vitro induction of unscheduled DNA synthesis (UDS) in primary hepatocytes of rat, mouse, hamster, rabbit and monkey. NC Blue No. 1, which is identified as a carcinogen by the National Toxicology Program, induced UDS in all 5 systems. HC Blue No. 2, which is identified as a non-carcinogen, induced UDS in rat, mouse, hamster and rabbit primary hepatocytes. 3-Methylcholanthrene and methyl methanesulfonate were used as positive controls to determine the sensitivity of the test system.