MilliporeSigma
  • Sensory nerves and nitric oxide contribute to reflex cutaneous vasodilation in humans.
Sensory nerves and nitric oxide contribute to reflex cutaneous vasodilation in humans.
American journal of physiology. Regulatory, integrative and comparative physiology (2013-02-15)
Brett J Wong
ABSTRACT
We tested the hypothesis that inhibition of cutaneous sensory nerves would attenuate reflex cutaneous vasodilation in response to an increase in core temperature. Nine subjects were equipped with four microdialysis fibers on the forearm. Two sites were treated with topical anesthetic EMLA cream for 120 min. Sensory nerve inhibition was verified by lack of sensation to a pinprick. Microdialysis fibers were randomly assigned as 1) lactated Ringer (control); 2) 10 mM nitro-L-arginine methyl ester (L-NAME) to inhibit nitric oxide synthase; 3) EMLA + lactated Ringer; and 4) EMLA + L-NAME. Laser-Doppler flowmetry was used as an index of skin blood flow, and blood pressure was measured via brachial auscultation. Subjects wore a water-perfused suit, and oral temperature was monitored as an index of core temperature. The suit was perfused with 50°C water to initiate whole body heat stress to raise oral temperature 0.8°C above baseline. Cutaneous vascular conductance (CVC) was calculated and normalized to maximal vasodilation (%CVC(max)). There was no difference in CVC between control and EMLA sites (67 ± 5 vs. 69 ± 6% CVC(max)), but the onset of vasodilation was delayed at EMLA compared with control sites. The L-NAME site was significantly attenuated compared with control and EMLA sites (45 ± 5% CVC(max); P < 0.01). Combined EMLA + L-NAME site (25 ± 6% CVC(max)) was attenuated compared with control and EMLA (P < 0.001) and L-NAME only (P < 0.01). These data suggest cutaneous sensory nerves contribute to reflex cutaneous vasodilation during the early, but not latter, stages of heat stress, and full expression of reflex cutaneous vasodilation requires functional sensory nerves and NOS.
Sigma-Aldrich
Prilocaine hydrochloride, ≥98% (TLC)