MilliporeSigma
  • Multi-responsive nitrobenzene-based amphiphilic random copolymer assemblies.
Multi-responsive nitrobenzene-based amphiphilic random copolymer assemblies.
Chemical communications (Cambridge, England) (2013-02-12)
Hao Wu, Jie Dong, Cancan Li, Yibiao Liu, Ning Feng, Liping Xu, Xiaowei Zhan, Huai Yang, Guojie Wang
ABSTRACT
A photo-, acid- and thermo-responsive nitrobenzene-based amphiphilic copolymer was synthesized and the corresponding multi-responsive behavior of the copolymer assemblies was revealed by TEM, DLS and AFM. The morphological changes of the assemblies under multi-stimuli endowed them with a controlled release of encapsulated molecules.
Product Description
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%