MilliporeSigma
  • Fate and removal of typical pharmaceuticals and personal care products by three different treatment processes.
Fate and removal of typical pharmaceuticals and personal care products by three different treatment processes.
The Science of the total environment (2013-02-07)
Yu-Jie He, Wei Chen, Xiao-Ying Zheng, Xing-Nan Wang, Xi Huang
ABSTRACT
The presence and distribution of typical of pharmaceuticals and personal care products (PPCPs), which comprise two types of polycyclic musks (PCMs) including Galaxolide (HHCB) and Tonalide (AHTN) as well as six types of estrogens containing estrone (E1), 17β-estradiol (E2), estriol (E3), 17α-ethynylestradiol (EE2), diethylstilbestrol (DES), and bisphenol A (BPA), were investigated at two wastewater treatment plants (WWTPs) in Jiangsu, China. Only raw wastewater was treated in WWTP A while WWTP B was serving an urban-industrialized area. In the influent, the concentrations of EE2 (2193-4437ngL(-1)), E2 (1126-1170ngL(-1)), and DES (268-421ngL(-1)) were generally higher than the previously reported values, whereas the concentrations of HHCB (306-316ngL(-1)), E1 (29-129ngL(-1)), E3 (53ngL(-1)), and BPA (26-176ngL(-1)) were much lower than those reported in other previous studies. In addition, AHTN was not detected in either WWTP and E3 was not found in WWTP B. The detected processes including anaerobic/oxic process (A/O), combined orbal oxidation ditch process (C-orbal OD) and anaerobic/anoxic/anoxic/oxic membrane biological reactor (A/A/A/O-MBR) showed higher removal efficiencies for HHCB (67-71%) and EE2 (87%) than those in other previous studies. Besides, the total hydraulic retention time (HRT) ranged between 6.7 and 20.0h, sludge retention time (SRT) ranged between 8 and 23d, and water temperature ranged from 24.8 to 28.2°C. The removal efficiencies for estrogens in biological processes were related to the following factors: the level of hydrophobic estrogens, the type of removal process (C-orbal OD was consistently less efficient in removing estrogens than A/O and A/A/A/O-MBR), and a high SRT or HRT (A/A/A/O-MBR with higher SRT and HRT showed higher and more stable removal of hydrophobic estrogens).