MilliporeSigma
Cardiovascular complications of Khat.
Clinica chimica acta; international journal of clinical chemistry (2013-02-02)
Jassim Al Suwaidi, Waleed M Ali, Samir L Aleryani
ABSTRACT
This article reviews current knowledge in khat (cathinone) research and its health impacts and toxicity in the cardiac system of khat chewers based on current evidence. The authors describe the process by which khat is postulated to induce multiple cardiac abnormalities by illustrating multiple case reports as described from around the world. The role of free radicals in inducing cardiac disease is also discussed. Future research areas are proposed to enhance our understanding of the mode of action of this toxicological drug.
Product Description
Supelco
R(+)-Cathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®