MilliporeSigma
  • Loop diuretic and anion modification of NEM-induced K transport in human red blood cells.
Loop diuretic and anion modification of NEM-induced K transport in human red blood cells.
The American journal of physiology (1990-04-01)
L R Berkowitz
ABSTRACT
The thioalkylating agent N-ethylmaleimide (NEM) causes ouabain-insensitive K loss from human red blood cells. This K loss is inhibited when intracellular Cl is replaced by another permeant anion or when loop diuretics are placed in the incubation medium after NEM exposure. In this report, we have tested the possibility that Cl replacement or loop diuretics not only influence the transport of K induced by NEM but also the interaction of NEM with its target sulfhydryl group. This possibility was examined by replacing intracellular Cl or exposing the cells to loop diuretics before NEM exposure, then measuring K loss in a Cl medium free of loop diuretics. We found that such pretreatment with either Cl substitution or loop diuretics stimulated, rather than inhibited, NEM-induced K loss. This enhancement was not additive in that the increase in K loss induced by anion substitution was not increased further when loop diuretics were also present. These data suggest that anion substitution and loop diuretics enhance the interaction of NEM with its cellular target but inhibit the K loss induced by NEM.
Sigma-Aldrich
Benzthiazide