MilliporeSigma
  • D-Alpha-tocopherol acid polyethylene glycol 1000 succinate, an effective stabilizer during solidification transformation of baicalin nanosuspensions.
D-Alpha-tocopherol acid polyethylene glycol 1000 succinate, an effective stabilizer during solidification transformation of baicalin nanosuspensions.
International journal of pharmaceutics (2013-01-08)
Peng-Fei Yue, Jing Wan, Yong Wang, Yu Li, Yue-Qin Ma, Ming Yang, Peng-Yi Hu, Hai-Long Yuan, Chang-Hong Wang
ABSTRACT
Baicalin nanosuspensions, stabilized with 10% TPGS (relative to the weight of baicalin), were transformed into nanosuspensions powders by solidification process. Solidification methods for this transformation included freeze-drying, spray drying or vacuum drying. High pressure homogenization was applied for production of baicalin nanosuspensions used TPGS, SDS, P188, HPMC and MC as stabilizer, respectively. The influence of the different solidification transformation methods on the redispersibility of solid drug nanosuspensions was systemically investigated, such as freeze-drying, spray drying and vacuum drying. Each method was applied with three grades of process stresses called as "conservative", "moderate" and "aggressive" conditions, and the redispersibility index (RDI) of nanosuspensions stabilized by stabilizers (such as TPGS, SDS, P188, HPMC and MC) during those process was investigated. The results showed that there was significant difference in RDI of nanosuspensions after solidification process. The RDI(a) (1.09, 1.01, 1.05, 0.99), RDI(b) (1.03, 0.99, 1.06, 1.02) and RDI(c) (1.01, 1.01, 1.09, 1.08) of nanosuspensions stabilized by TPGS were more small during different solidification process, compared with those of nanosuspensions stabilized by other stabilizer. It was concluded that the baicalin nanosuspensions were subjected to agglomeration or crystal growth during solidification transformation, especially at high aggressive stress conditions. Meanwhile, compared to other stabilizer, the TPGS was more effective for stability of baicalin nanosuspensions, which could exhibit higher affinity to the drug crystal and stronger surface adsorption at different solidification stresses.
Sigma-Aldrich
Baicalin, 95%