MilliporeSigma
  • An integrated assessment of sediment remediation in a midwestern U.S. stream using sediment chemistry, water quality, bioassessment, and fish biomarkers.
An integrated assessment of sediment remediation in a midwestern U.S. stream using sediment chemistry, water quality, bioassessment, and fish biomarkers.
Environmental toxicology and chemistry (2012-12-13)
John R Meier, Steve Snyder, Victoria Sigler, Dave Altfater, Mike Gray, Bill Batin, Paul Baumann, Denise Gordon, Paul Wernsing, Jim Lazorchak
ABSTRACT
A comprehensive biological, sediment, and water quality study of the lower Little Scioto River near Marion, Ohio, USA, was undertaken to evaluate the changes or improvements in biotic measurements following the removal of creosote-contaminated sediment. The study area covered 7.5 river miles (RMs), including a remediated section between RMs 6.0 and 6.8. Fish and macroinvertebrate assemblages, fish biomarkers (i.e., polycyclic aromatic hydrocarbon [PAH] metabolite levels in white sucker [Castostomus commersoni] and common carp [Cyprinus carpio] bile and DNA damage), sediment chemistry, and water quality were assessed at five locations relative to the primary source of historical PAH contamination-upstream (RM 9.2), adjacent (RM 6.5), and downstream (RMs 5.7, 4.4, and 2.7). Overall, the biomarker results were consistent with the sediment PAH results, showing a pattern of low levels of PAH bile metabolites and DNA damage at the upstream (reference or background location), as well as the remediated section, high levels at the two immediate downstream sites, and somewhat lower levels at the furthest downstream site. Results show that remediation was effective in reducing sediment contaminant concentrations and exposure of fish to PAHs and in improving fish assemblages (60% increase in index of biotic integrity scores) in remediated river sections. Additional remedial investigation and potentially further remediation is needed to improve the downstream benthic fish community, which is still heavily exposed to PAH contaminants.
Sigma-Aldrich
Creosote from beechwood tar