MilliporeSigma
Timed insemination of beef heifers using the 7-11 Synch protocol.
Journal of animal science (2012-12-12)
D R Eborn, D M Grieger
ABSTRACT
Two experiments were conducted over 3 yr to determine pregnancy rates in beef heifers after a timed AI in response to the 7-11 Synch protocol. In Exp. 1, 179 heifers were either fed melengestrol acetate (MGA; 7-11 Synch) or given an intravaginal progesterone (P4)-releasing insert [controlled intravaginal drug releasing device (CIDR); 7-11 CIDR] for 7 d. Prostaglandin F2αwas administered on the last day of MGA feeding or at CIDR removal followed by the CO-Synch protocol (GnRH-PGF2α-GnRH) beginning 4 d after MGA withdrawal or 2 d after CIDR removal. Heifers received a timed AI with GnRH beginning 48 h after the second PGF2α. Blood samples were collected at d -10, 0 (start of MGA feeding), and 18 (second PGF2α injection). In Exp. 2, 298 beef heifers were treated with the 7-11 Synch protocol with (7-11 Synch) or without (7 Synch) the first GnRH injection. Fixed time AI and GnRH was given 54 h after PGF2α. Blood samples were collected at d -10 and 0 in yr 1 and d -10, 0, 18 and at AI in yr 2. In Exp. 1, no differences were detected between 7 and 11 Synch and 7-11 CIDR for attainment of puberty in noncyclic heifers (94 vs. 78%; P = 0.21), the proportion of heifers that had luteal tissue on d 18 (87 vs. 83%; P = 0.41), or pregnancy rates after timed AI (47 vs. 46%; P = 0.99). In Exp. 2, administration of GnRH 4 d after the last MGA (7-11 Synch) feeding tended (P = 0.07) to induce more prepubertal heifers to cycle (88 vs. 61%) and increased (P < 0.01) the proportion of heifers with luteal tissue on d 18 (88 vs. 72%) compared with heifers in the 7 Synch treatment. Pregnancy rates after the 54 h timed AI were greater (P < 0.01) in the 7-11 Synch treatment (55%) than in the 7 Synch (38%). We conclude that heifer pregnancy rates did not differ whether feeding MGA for 7 d or applying a CIDR insert for 7 d before a CO-Synch protocol. In contrast, use of GnRH at the beginning of the CO-Synch protocol improved pregnancy rates after a timed AI by inducing more prepubertal heifers to ovulate and increasing the proportion of heifers with luteal tissue at the PGF2α injection.
Product Description
Supelco
Melengestrol acetate, VETRANAL®, analytical standard