MilliporeSigma
  • Postoperative subcutaneous instillation of low-dose ketorolac but not hydromorphone reduces wound exudate concentrations of interleukin-6 and interleukin-10 and improves analgesia following cesarean delivery.
Postoperative subcutaneous instillation of low-dose ketorolac but not hydromorphone reduces wound exudate concentrations of interleukin-6 and interleukin-10 and improves analgesia following cesarean delivery.
The journal of pain : official journal of the American Pain Society (2012-12-12)
Brendan Carvalho, Harry J Lemmens, Vicki Ting, Martin S Angst
ABSTRACT
The objectives of this study were to test the effects of low-dose ketorolac and hydromorphone added to continuous local anesthetic wound instillation on surgical-site inflammatory mediators, postoperative pain, and opioid consumption. Sixty healthy women undergoing cesarean delivery were enrolled in this randomized, double-blinded study. Patients were randomized to receive a subcutaneous wound instillation of bupivacaine .5% at 10 mg/hour (active control), bupivacaine .5% with ketorolac .6 mg/hour, or bupivacaine .5% with hydromorphone .04 mg/hour for 48 hours postcesarean. Wound exudate was sampled at 4, 24, and 48 hours postcesarean and assayed for interleukins IL-1β, IL-2, IL-6, IL-8, IL-10, and IL-12, tumor necrosis factor (TNF-α), interferon (INF-γ), and granulocyte-macrophage colony stimulating factor (GM-CSF). The addition of ketorolac to bupivacaine significantly decreased IL-6 (P = .012) and IL-10 (P = .005) compared to plain bupivacaine. Ketorolac, but not hydromorphone, was associated with a decrease in pain (P = .018) and analgesic use (P = .020) following cesarean delivery. Our results are compatible with the view that significant analgesics effects are mediated through local modulation of inflammatory events. Low-dose ketorolac administered into surgical wounds exert significant anti-inflammatory and analgesic effects and may be a valuable analgesic alternative to systemic nonsteroidal anti-inflammatories (NSAIDs) but with potentially fewer side effects. This article demonstrates that low-dose ketorolac administered into wounds modulates local inflammatory events, decreases postoperative pain, and reduces opioid consumption. These results suggest that administration of NSAIDs into surgical wounds may be an analgesic alternative to higher systemic dosing of NSAIDs.
Product Description
Supelco
Hydromorphone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®