MilliporeSigma
  • Adhesion-preventing properties of 4% icodextrin and canola oil: a comparative experimental study.
Adhesion-preventing properties of 4% icodextrin and canola oil: a comparative experimental study.
Clinics (Sao Paulo, Brazil) (2012-11-28)
Cengizhan Yigitler, Dursun Ozgur Karakas, Zafer Kucukodaci, Alpaslan Cosar, Bülent Gülec, Mehmet Levhi Akin
ABSTRACT
Postsurgical abdominal adhesions are common, serious postoperative complications. The present study compared the usefulness of 4% icodextrin and canola oil in preventing postoperative peritoneal adhesions. Twenty-four Wistar albino rats were divided into three groups. Following a laparotomy, a serosal abrasion was made by brushing the cecum, and 3 mL of 0.9% NaCl, 4% icodextrin, or 3 mL of canola oil were intraperitoneally administered for the control, icodextrin, and canola oil groups, respectively. The abdomen was then closed. All of the rats were sacrificed at day 10. Macroscopic, histopathological, and biochemical evaluations were performed. The results were statistically analyzed using Kruskal-Wallis and ANOVA tests. Macroscopic analyses revealed that both canola oil and 4% icodextrin reduced adhesion formation, but the difference was not statistically significant (p = 0.17). The histopathological examinations revealed no significant differences in terms of giant cell, lymphocyte/plasmocyte, neutrophil, ICAM1, or PECAM1 scores. However, both canola oil and 4% icodextrin significantly reduced fibrosis (p = 0.025). In the canola oil group, the histiocytic reactions were significantly increased (p = 0.001), and the hydroxyproline levels were significantly lower than those in the other groups (p = 0.034). In the present study, canola oil was determined to be superior to 4% icodextrin in lowering hydroxyproline levels and increasing histiocytic reactions. Considering these results, we believe that canola oil is a promising agent for preventing adhesion formation.