MilliporeSigma
  • Synthesis of (-)-morphine: application of sequential Claisen/Claisen rearrangement of an allylic vicinal diol.
Synthesis of (-)-morphine: application of sequential Claisen/Claisen rearrangement of an allylic vicinal diol.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-11-28)
Masato Ichiki, Hiroki Tanimoto, Shohei Miwa, Ryosuke Saito, Takaaki Sato, Noritaka Chida
ABSTRACT
A detailed exploration of the synthesis of (-)-morphine based on sequential [3,3]-sigmatropic rearrangements is described. The sequential Claisen/Claisen rearrangements of an allylic vicinal diol resulted in the stereoselective formation of the two contiguous carbon centers, including a sterically encumbered quaternary carbon, in a single operation. The two ethyl esters generated in this reaction were successfully differentiated during a subsequent Friedel-Crafts-type cyclization. The (-)-morphine double bond was introduced at a late stage in our first-generation synthesis, but was formed at an earlier stage in the second-generation synthesis, resulting in a more efficient route to the end product.
Sigma-Aldrich
Allyl alcohol, ≥99%
Sigma-Aldrich
Allyl alcohol, ≥98.5%