MilliporeSigma
Trifluoromethylation of alkenes by visible light photoredox catalysis.
The Journal of organic chemistry (2012-11-22)
Naeem Iqbal, Sungkyu Choi, Eunjin Kim, Eun Jin Cho
ABSTRACT
A method for trifluoromethylation of alkenes has been developed employing visible light photoredox catalysis with CF(3)I, Ru(Phen)(3)Cl(2), and DBU. This process works especially well for terminal alkenes to give alkenyl-CF(3) products with only E-stereochemistry. The mild reaction conditions enable the trifluoromethylation of a range of alkenes that bear various functional groups.