MilliporeSigma
  • Enantioselective analysis of zopiclone in rat brain by liquid chromatography tandem mass spectrometry.
Enantioselective analysis of zopiclone in rat brain by liquid chromatography tandem mass spectrometry.
Analytical and bioanalytical chemistry (2012-11-07)
Milena Araújo Tonon, Valquíria A P Jabor, Pierina Sueli Bonato
ABSTRACT
A new high-performance liquid chromatographic method with triple quadrupole mass spectrometry detection was developed and validated for the quantification of zopiclone enantiomers in rat brain samples. Zopiclone enantiomers were resolved on a CHIRALPAK AD column with a mobile phase consisting of acetonitrile/ethanol/methanol (60:20:20, v/v/v) at a flow rate of 1.3 mL min(-1). Moclobemide was used as internal standard. The sample treatment procedure was carried out employing solid-phase extraction, yielding mean absolute recoveries of 89.6 and 91.7% for each zopiclone enantiomer. The validated method showed linearity in the range of 0.29-344.8 ng g(-1), with quantification limits of 0.29 ng g(-1) for both enantiomers. Precision and accuracy were within acceptable levels of confidence (<15%). The method was applied in a pilot study of zopiclone kinetic disposition in rats. It could be observed that the levels of (+)-(S)-zopiclone were always higher than those of (-)-(R)-zopiclone, confirming the stereoselective disposition of zopiclone.
Sigma-Aldrich
Zopiclone
Supelco
Zopiclone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®