MilliporeSigma
Discriminative stimulus effects of N,N-diisopropyltryptamine.
Psychopharmacology (2012-10-17)
Theresa M Carbonaro, Michael J Forster, Michael B Gatch
ABSTRACT
Serotonergic hallucinogens such as (+)-lysergic acid diethylamide (LSD) and dimethyltryptamine (DMT) produce distinctive visual effects, whereas the synthetic hallucinogen N,N-diisopropyltryptamine (DiPT) is known for its production of auditory distortions. This study compares the discriminative stimulus effects of DiPT to those of visual hallucinogens. Adult male rats were trained to discriminate DiPT (5 mg/kg, 15 min) from saline under a FR10 schedule. A dose-effect and time course of DiPT's discriminative stimulus effects were established. DMT, (-)-2,5-dimethoxy-4-methylamphetamine (DOM), LSD, (±)-methylenedioxymethamphetamine (MDMA), and (+)-methamphetamine were tested for cross-substitution in DiPT-trained animals. Rats learned to discriminate DiPT from saline in an average of 60 training sessions (30 drug and 30 saline). DiPT (0.5-5 mg/kg) produced dose-dependent increases in drug-appropriate responding (DAR) to 99 % (ED(50) = 2.47 mg/kg). Onset of the discriminative stimulus effects was within 5 min, and the effects dissipated within 4 h. Full substitution for the discriminative stimulus effects of DiPT occurred with LSD, DOM, and MDMA. DMT only partially substituted for DiPT (65 % DAR), whereas (+)-methamphetamine failed to substitute for DiPT (29 % DAR). The discriminative stimulus effects of DiPT were similar those of a number of synthetic hallucinogens, only partially similar to those of DMT, but not similar to (+)-methamphetamine. The putative DiPT-induced auditory distortions do not lead to discriminative stimulus effects distinguishable from other hallucinogens.
Product Description
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®