MilliporeSigma
  • Cu(GlyGlyHis) effects on MCF7 cells: copper uptake, reactive oxygen species generation and membrane topography changes.
Cu(GlyGlyHis) effects on MCF7 cells: copper uptake, reactive oxygen species generation and membrane topography changes.
Journal of inorganic biochemistry (2012-10-02)
Andreza C Matias, Nathália Villa dos Santos, Rodrigo Chelegão, Cassiana S Nomura, Pablo A Fiorito, Giselle Cerchiaro
ABSTRACT
Elevated levels of copper have been detected in various types of human cancer cells, such as breast cancer cells, and a number of mechanisms have been proposed to explain the action and influence of copper on tumor progress. In this work, we found that stimulating the proliferation of mammary epithelial MCF7 cells with the high-redox-potential copper complex Cu (GlyGlyHis) is associated with the copper-induced intracellular generation of reactive oxygen species (ROS) that induces lipid peroxidation and causes increased roughness of external cell membranes, which leads to the formation of larger cell domes. The results presented herein provide new insights into the molecular link between copper and the proliferation of breast cancer cells and, consequently, into the mechanism by which changes in redox balance and ROS accumulation regulates cell membrane roughness.
Sigma-Aldrich
Gly-Gly-His