MilliporeSigma
  • A randomized controlled non-inferiority study comparing the antiemetic effect between intravenous granisetron and oral azasetron based on estimated 5-HT3 receptor occupancy.
A randomized controlled non-inferiority study comparing the antiemetic effect between intravenous granisetron and oral azasetron based on estimated 5-HT3 receptor occupancy.
Anticancer research (2012-09-21)
Junki Endo, Hirotoshi Iihara, Maya Yamada, Koumei Yanase, Fumihiko Kamiya, Fumitaka Ito, Norihiko Funaguchi, Yasushi Ohno, Shinya Minatoguchi, Yoshinori Itoh
ABSTRACT
The acute antiemetic effect was compared between oral azasetron and intravenous granisetron based on the 5-hydroxytryptamine(3) (5-HT(3)) receptor occupancy theory. Receptor occupancy was estimated from reported data on plasma concentrations and affinity constants to 5-HT(3) receptor. A randomized non-inferiority study comparing acute antiemetic effects between oral azasetron and intravenous granisetron was performed in 105 patients receiving the first course of carboplatin-based chemotherapy for lung cancer. Azasetron exhibited the highest 5-HT(3) receptor occupancy among various first-generation 5-HT(3) antagonists. The complete response to oral azasetron was shown to be non-inferior to that of intravenous granisetron, in which the risk difference was 0.0004 (95% confidence interval: -0.0519-0.0527). The lower limit of the confidence intervals did not exceed the negative non-inferiority margin (-0.1). The complete response during the overall period was not different (68% versus 67%). Oral azasetron was found to be non-inferior to intravenous granisetron in the acute antiemetic effect against moderately emetogenic chemotherapy.
Sigma-Aldrich
Granisetron hydrochloride, ≥98% (HPLC), solid
Granisetron hydrochloride, European Pharmacopoeia (EP) Reference Standard