MilliporeSigma
  • Anomalous size-dependent decay of low-energy luminescence from PbS quantum dots in colloidal solution.
Anomalous size-dependent decay of low-energy luminescence from PbS quantum dots in colloidal solution.
ACS nano (2012-09-14)
Elena V Ushakova, Aleksandr P Litvin, Peter S Parfenov, Anatoly V Fedorov, Mikhail Artemyev, Anatoly V Prudnikau, Ivan D Rukhlenko, Alexander V Baranov
ABSTRACT
We report on an anomalous size dependence of the room-temperature photoluminescence decay time from the lowest-energy state of PbS quantum dots in colloidal solution, which was found using the transient luminescence spectroscopy. The observed 10-fold reduction in the decay time (from ~2.5 to 0.25 μs) with the increase in the quantum dots' diameter is explained by the existence of phonon-induced transitions between the in-gap state-whose energy drastically depends on the diameter-and the fundamental state of the quantum dots.