MilliporeSigma
  • N-methylated tryptamine derivatives in citrus genus plants: identification of N,N,N-trimethyltryptamine in bergamot.
N-methylated tryptamine derivatives in citrus genus plants: identification of N,N,N-trimethyltryptamine in bergamot.
Journal of agricultural and food chemistry (2012-09-11)
Luigi Servillo, Alfonso Giovane, Maria Luisa Balestrieri, Domenico Cautela, Domenico Castaldo
ABSTRACT
The occurrence of N-methylated tryptamine derivatives in bergamot plant (Citrus bergamia Risso et Poit) is reported for the first time. Interestingly, the most abundant of these substances is N,N,N-trimethyltryptamine, which has not been previously identified in any citrus plant. The N-methylated tryptamine derivatives were identified and quantitated in leaves, peel, juice, and seeds by high-performance liquid chromatography-electrospray ionization-tandem mass spectrometry. N,N,N-Trimethyltryptamine was confirmed by MS(3) and comparison with the synthesized authentic standard. In addition, the study of the distribution of tryptophan, tryptamine, N-methyltryptamine, N,N-dimethyltryptamine, and N,N,N-trimethyltryptamine indicated that these compounds are differently expressed in the various tissues of the bergamot plant. Intriguingly, chemically synthesized N,N,N-trimethyltryptamine was reported to possess nicotine-like activity being a stimulant of parasympathetic ganglia by exerting its action on acetylcholine receptors. On this basis, the identification of N,N,N-trimethyltryptamine at a relatively high level in leaves suggests a possible role in a physiological mechanism of plant defense.
Sigma-Aldrich
N-ω-Methyltryptamine, 99%