MilliporeSigma
  • Attenuated effect of tungsten carbide nanoparticles on voltage-gated sodium current of hippocampal CA1 pyramidal neurons.
Attenuated effect of tungsten carbide nanoparticles on voltage-gated sodium current of hippocampal CA1 pyramidal neurons.
Toxicology in vitro : an international journal published in association with BIBRA (2012-09-08)
Dehong Shan, Yongling Xie, Guogang Ren, Zhuo Yang
ABSTRACT
Nanomaterials and relevant products are now being widely used in the world, and their safety becomes a great concern for the general public. Tungsten carbide nanoparticles (nano-WC) are widely used in metallurgy, aeronautics and astronautics, however our knowledge regarding the influence of nano-WC on neurons is still lacking. The aim of this study was to investigate the impact of nano-WC on tetrodotoxin (TTX)-sensitive voltage-activated sodium current (I(Na)) of hippocampal CA1 pyramidal neurons. Results showed that acute exposure of nano-WC attenuated the peak amplitudes of I(Na) in a concentration-dependent manner. The minimal effective concentration was 10(-5)g/ml. The exposure of nano-WC significantly decreased current amplitudes of the current-voltage curves of I(Na) from -50 to+50 mV, shifted the steady-state activation and inactivation curves of I(Na) negatively and delayed the recovery of I(Na) from inactivation state. After exposure to nano-WC, the peak amplitudes, overshoots and the V-thresholds of action potentials (APs) were markedly reduced. These results suggested that exposure of nano-WC could influence some characteristics of APs evoked from the hippocampal CA1 neurons by modifying the kinetics of voltage-gated sodium channels (VGSCs).
Sigma-Aldrich
Tungsten(IV) carbide, powder, 2 μm, ≥99%