MilliporeSigma
  • Analyzing diurnal and age-related pheromone emission of the olive fruit fly, Bactrocera oleae by sequential SPME-GCMS analysis.
Analyzing diurnal and age-related pheromone emission of the olive fruit fly, Bactrocera oleae by sequential SPME-GCMS analysis.
Journal of chemical ecology (2012-09-04)
Anat Levi-Zada, David Nestel, Daniela Fefer, Esther Nemni-Lavy, Inbal Deloya-Kahane, Maayan David
ABSTRACT
The olive fruit fly, Bactrocera oleae (Diptera: Tephritidae), uses 1,7-dioxaspiro[5.5]undecane ("olean"), produced primarily by females, as a sex pheromone. We used sequential solid phase microextraction-gas chromatography mass spectrometry (SPME-GCMS) analysis to show that female olive flies release about 1000 ng of pheromone at the onset of scotophase for several weeks, while males release about 1/100 as much during the first week after eclosion. The present research demonstrates details of employing SPME-GCMS with the partially known pheromone system of the olive fruit fly as a model for pheromone identification and diurnal release patterns in insects, especially fruit flies. The sequential SPME-GCMS method will readily allow detection and semi-quantification of semiochemicals released by insects in minute amounts throughout the diurnal cycle.
Sigma-Aldrich
1,7-Dioxaspiro[5.5]undecane, ≥97%