MilliporeSigma
  • A survey of the FDA's AERS database regarding muscle and tendon adverse events linked to the statin drug class.
A survey of the FDA's AERS database regarding muscle and tendon adverse events linked to the statin drug class.
PloS one (2012-09-01)
Keith B Hoffman, Christina Kraus, Mo Dimbil, Beatrice A Golomb
ABSTRACT
Cholesterol management drugs known as statins are widely used and often well tolerated; however, a variety of muscle-related side effects can arise. These adverse events (AEs) can have serious impact, and form a significant barrier to therapy adherence. Surveillance of post-marketing AEs is of vital importance to understand real-world AEs and reporting differences between individual statin drugs. We conducted a review of post-approval muscle and tendon AE reports in association with statin use, to assess differences within the drug class. We analyzed all case reports from the FDA AE Reporting System (AERS) database linking muscle-related AEs to statin use (07/01/2005-03/31/2011). Drugs examined were: atorvastatin, simvastatin, lovastatin, pravastatin, rosuvastatin, and fluvastatin. Relative risk rates for rosuvastatin were consistently higher than other statins. Atorvastatin and simvastatin showed intermediate risks, while pravastatin and lovastatin appeared to have the lowest risk rates. Relative risk of muscle-related AEs, therefore, approximately tracked with per milligram LDL-lowering potency, with fluvastatin an apparent exception. Incorporating all muscle categories, rates for atorvastatin, simvastatin, pravastatin, and lovastatin were, respectively, 55%, 26%, 17%, and 7.5% as high, as rosuvastatin, approximately tracking per milligram potency (Rosuvastatin>Atorvastatin>Simvastatin>Pravastatin ≈ Lovastatin) and comporting with findings of other studies. Relative potency, therefore, appears to be a fundamental predictor of muscle-related AE risk, with fluvastatin, the least potent statin, an apparent exception (risk 74% vs rosuvastatin). AE reporting rates differed strikingly for drugs within the statin class, with relative reporting aligning substantially with potency. The data presented in this report offer important reference points for the selection of statins for cholesterol management in general and, especially, for the rechallenge of patients who have experienced muscle-related AEs (for whom agents of lower expected potency should be preferred).
Product Description
Fluvastatin sodium, European Pharmacopoeia (EP) Reference Standard
Fluvastatin for system suitability, European Pharmacopoeia (EP) Reference Standard