MilliporeSigma
  • [Carbon disulfide exposure level of workers in a chemical fiber industry].
[Carbon disulfide exposure level of workers in a chemical fiber industry].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2012-08-31)
Kui-rong Li, Shou-ming Cui, Hui Wu, Li-min Guo, Jun-ying Ma, Gui-zhen Gu, Shan-fa Yu
ABSTRACT
To investigate the exposure levels of carbon disulfide (CS(2)) for a chemical fiber industry. The concentration of CS(2) was monitored in representative workshops and types of work, and the datas of that over the years were collected. The short-term exposure concentration of CS(2) about 80% of the type of work was less than or equal to 10 mg/m(3), which of more than 90% was less than or equal to 20 mg/m(3). The time weighted average concentration of CS(2) about 70% of the type of work was less than or equal to 5 mg/m(3), which of more than 90% was less than or equal to 10 mg/m(3). The short-term exposure concentration of CS(2) which was more than 15 mg/m(3) or the time weighted average concentration of CS(2) which was more than 30 mg/m(3) was only for little type of work. The concentration of CS(2) for the most type of work was lower, but there were still a number of types of work exposuring the higher concentration, which exceed the national occupational exposure limits.
Product Description
Supelco
Carbon disulfide solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Carbon disulfide, anhydrous, ≥99%
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, low benzene, ≥99.9%
Sigma-Aldrich
Carbon disulfide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, purified by redistillation, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Sigma-Aldrich
Carbon disulfide, suitable for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC)