MilliporeSigma
  • Asymmetric syntheses of (-)-pentazocine and (-)-eptazocine through an aza-Prins cyclization.
Asymmetric syntheses of (-)-pentazocine and (-)-eptazocine through an aza-Prins cyclization.
Chemistry, an Asian journal (2012-08-22)
Qiang Chen, Xing Huo, Zhen Yang, Xuegong She
ABSTRACT
Two down more to go: The asymmetric syntheses of (-)-pentazocine and (-)-eptazocine are presented. The highlights of the syntheses are the construction of the core skeleton through an aza-Prins cyclization and intramolecular Friedel-Crafts reaction.
Product Description
Supelco
Pentazocine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®