MilliporeSigma
  • First enantioselective total synthesis of (+)-(R)-Pinnatolide using an asymmetric domino allylation reaction.
First enantioselective total synthesis of (+)-(R)-Pinnatolide using an asymmetric domino allylation reaction.
Organic letters (2012-08-03)
Lutz F Tietze, Thomas Wolfram, Julian J Holstein, Birger Dittrich
ABSTRACT
An efficient total synthesis of (+)-(R)-Pinnatolide is described. As a key step an asymmetric multicomponent domino allylation reaction of methyl levulinate is used to form the quaternary stereogenic center in a highly selective way.
Sigma-Aldrich
Methyl levulinate, ≥98.0%