MilliporeSigma
  • Monitoring of methylergometrine in human breast milk by solid-phase extraction and high-performance liquid chromatography with fluorimetric detection.
Monitoring of methylergometrine in human breast milk by solid-phase extraction and high-performance liquid chromatography with fluorimetric detection.
Die Pharmazie (2012-07-25)
T Nakamichi, A Yawata, H Hojo, H Kagaya, S Kobayashi, T Chikuma
ABSTRACT
A high-performance liquid chromatographic assay has been developed for the detection and quantification of the conventional postnatal uterotonic drug, methylergometrine, in human breast milk using a C-18 reversed-phase column by isocratic elution. The analytical method consisted of sample clean-up by solid-phase extraction, and the fluorescence detection required only 8.5 min per sample for separation and quantitation. This assay gave intra- and inter-assay coefficients of variation of less than 7.9% and 7.7%, respectively, and the detection limit was approximately 50 pg/ml. This method was applied for drug level monitoring in the breast milk of patients given methylergometrine.