MilliporeSigma
  • Chiral analysis of methadone and its main metabolite, EDDP, in postmortem brain and blood by automated SPE and liquid chromatography-mass spectrometry.
Chiral analysis of methadone and its main metabolite, EDDP, in postmortem brain and blood by automated SPE and liquid chromatography-mass spectrometry.
Journal of analytical toxicology (2012-07-11)
Karen Marie Dollerup Holm, Kristian Linnet
ABSTRACT
We developed a method based on liquid chromatography coupled with tandem mass spectrometry to quantify individual enantiomers of methadone and its primary metabolite, R/S-2-ethyl-1,5-dimethyl-3,3-diphenylpyrrolinium (EDDP), in postmortem blood and brain tissue. Samples were prepared with a Tecan Evo robotic system. Precipitation was followed by solid-phase extraction, evaporation and reconstitution in the mobile phase. Enantiomers were fully separated with liquid chromatography on a chiral α(1)-acid glycoprotein column. A Quattro micro mass spectrometer was used for detection in the positive ion mode with an electrospray source. The lower limit of quantification in brain tissue was 0.005 mg/kg for methadone and 0.001 mg/kg for EDDP enantiomers; the maximum precision was 17% for both compounds; accuracy ranged from 94 to 101%. In blood, the limit of quantification was 0.001 mg/kg for all compounds, the total relative standard deviation was <15%, and the accuracy varied from 95 to 109%. Brain (n = 11) and blood (n = 15) samples were analyzed with intermediate precision that varied from 7.5 to 15% at 0.005 mg/kg and from 6.8 to 11.3% at 0.25 mg/kg for all compounds. Method development focused on producing a clean extract, particularly from brain samples. The method was tested on authentic brain and femoral blood samples.
Product Description
Supelco
EDDP perchlorate solution, 1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®