MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Alantolactone induces activation of apoptosis in human hepatoma cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-06-23)
Jia-Chuan Lei, Jian-Qing Yu, Yan Yin, Yan-Wen Liu, Guo-Lin Zou
ABSTRACT
Alantolactone, a sesquiterpene lactone, possesses anti-inflammatory property. In this study, we provide evidence that it could be developed as a novel agent against human liver cancer. We observed that alantolactone treatment to HepG2, Bel-7402 and SMMC-7721 cells, human liver cancer cell lines resulted in a dose-dependent inhibition of cell growth. We selected HepG2 cell line as a test model system. Alantolactone treatment of HepG2 cells resulted in a dose-dependent induction of apoptosis and arrest of cells in G2-M phase. This induction of apoptosis seems to be mediated via modulating the protein levels of Bcl-2 family and activation of caspases. Moreover, caspase-8 and Bid activation, loss of mitochondrial transmembrane potential and cytochrome c release suggest the existence of a cross-talk between the death receptor and the mitochondrial pathways. We also observed that alantolactone treatment of cells resulted in a dose-dependent decrease in NF- κB/p65. In addition, a significant and progressive increase in the level of p53 protein in alantolactone-treated cells was observed. Taken together, our data suggest that alantolactone could be developed as an agent against human liver cancer.
Sigma-Aldrich
Alantolactone, ≥98% (HPLC)