MilliporeSigma
  • Mediating effects of aryl-hydrocarbon receptor and RhoA in altering brain vascular integrity: the therapeutic potential of statins.
Mediating effects of aryl-hydrocarbon receptor and RhoA in altering brain vascular integrity: the therapeutic potential of statins.
The American journal of pathology (2012-06-23)
Chih-Cheng Chang, Pei-Shan Lee, Ying Chou, Ling-Ling Hwang, Shu-Hui Juan
ABSTRACT
We have demonstrated previously that focal adhesion kinase (FAK)/RhoA alteration by the aryl-hydrocarbon receptor (AhR) agonist 3-methylcholanthrene (3MC) is involved in the antimigratory effects of 3MC in human umbilical vascular endothelial cells. Here, we identified that signaling properties and molecular mechanisms of RhoA/β-catenin were both implicated in alterations to blood-brain barrier integrity. The mechanisms of action were the down-regulation of integrin, the extracellular matrix, and adherens junction stability. PTEN phosphorylation by 3MC-mediated AhR/RhoA activation increased the proteasomal degradation of β-catenin through PKCδ/pGSK3β-mediated β-catenin phosphorylation; the crucial roles of AhR/RhoA in this process were verified by using gain- or loss-of-function experiments. The decrease in β-catenin led to decreased expression of fibronectin and α5β1 integrin. Additionally, protein interactions among FAK, VE-cadherin, vinculin, and β-actin were simultaneously decreased, resulting in adherens junction instability. Novel functional TCF/LEF1 binding sites in the promoter regions of fibronectin and α5/β1 integrin were identified by electrophoretic mobility shift and chromatin immunoprecipitation assays. The results indicate that the binding activities of β-catenin decreased in mouse cerebrovascular endothelial cells treated with 3MC. In addition, simvastatin and pravastatin treatment reversed 3MC-mediated alterations in mouse cerebrovascular endothelial cells by RhoA inactivation, and the in vitro findings were substantiated by an in vivo blood-brain barrier assay. Thus, endothelial barrier dysfunction due to 3MC occurs through AhR/RhoA-mediated β-catenin down-regulation, which is reversed by simvastatin treatment in vivo.
Product Description
Supelco
3-Methylcholanthrene solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
3-Methylcholanthrene, 98%
Supelco
3-Methylcholanthrene, analytical standard