MilliporeSigma
  • Analysis of lysergic acid amide in human serum and urine after ingestion of Argyreia nervosa seeds.
Analysis of lysergic acid amide in human serum and urine after ingestion of Argyreia nervosa seeds.
Analytical and bioanalytical chemistry (2012-06-15)
Alexander Paulke, Christian Kremer, Cora Wunder, Stefan W Toennes
ABSTRACT
The ergot alkaloid lysergic acid amide (LSA) is a secondary plant constituent in a number of plants, but it is mainly present in considerable amounts in Convolvulaceae, like Argyreia nervosa. Due to its close structural similarity to lysergic acid diethylamide, LSA is considered as psychedelic and therefore promoted as so-called "legal high" in various internet forums. During a human behavioral study with orally administered seeds of A. nervosa, blood and urine samples were obtained. The present study describes the validation of a sensitive and robust high performance liquid chromatography method with fluorescence detection, which was applied to the study samples. The limit of detection (LOD) and lower limit of quantification in human serum were 0.05 and 0.17 ng/mL, respectively, and in urine, the LOD was 0.15 ng/mL. Intra- and interday precision and accuracy were below 15 % relative standard deviation with a bias better than ±15 %. No conversion of LSA to its epimer iso-LSA was noted during analyses. The LSA concentrations in the authentic human serum samples were in the range of 0.66 to 3.15 ng/mL approximately 2 h after ingestion. In urine, LSA could be found 1-24 h after ingestion; after 48 h, no LSA could be detected. The LSA epimer iso-LSA was also detected in serum and urine in varying ratios. In conclusion, LSA serum levels in the low nanogram per milliliter range correlated with severe vegetative adverse effects (nausea, weakness, fatigue, tremor, blood pressure elevation) and a psychosis-like state, which led to study termination.
Product Description
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®