MilliporeSigma
  • Calciate-mediated intermolecular hydroamination of diphenylbutadiyne with secondary anilines.
Calciate-mediated intermolecular hydroamination of diphenylbutadiyne with secondary anilines.
Chemical communications (Cambridge, England) (2012-06-12)
Carsten Glock, Helmar Görls, Matthias Westerhausen
ABSTRACT
Calciate-mediated intermolecular hydroamination of diphenylbutadiyne with N-phenyl and N-isopropyl-substituted anilines yields E- and Z-isomers of the corresponding 1-anilino-1,4-diphenylbut-1-ene-3-yne. In the case of HNPh(2) solely heterobimetallic K(2)Ca(NPh(2))(4) is able to effectively catalyze this hydroamination reaction in tetrahydrofuran at elevated temperatures.