MilliporeSigma
  • Spectroscopic (infrared, Raman, UV and NMR) analysis, Gaussian hybrid computational investigation (MEP maps/HOMO and LUMO) on cyclohexanone oxime.
Spectroscopic (infrared, Raman, UV and NMR) analysis, Gaussian hybrid computational investigation (MEP maps/HOMO and LUMO) on cyclohexanone oxime.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-06-12)
S Ramalingam, M Karabacak, S Periandy, N Puviarasan, D Tanuja
ABSTRACT
In the present analysis, FT-IR/FT-Raman spectra of the cyclohexanone oxime (CHO, C(6)H(11)NO) are recorded. The observed vibrational frequencies are assigned and the computational calculations are carried out by HF and DFT (B3LYP and B3PW91) methods with 6-311++G(d,p) basis set and the corresponding results are tabulated. In order to yield good coherence with observed values, the calculated frequencies are scaled by appropriate scale factors. The complete assignments are performed on the basis of the total energy distribution (TED) of the vibrational modes, calculated with scaled quantum mechanics (SQM) method. The alternation of structure of cyclohexanone due to the substitution of NOH is investigated. The vibrational sequence pattern of the molecule related to the substitutions is analyzed. Comparison of the observed fundamental vibrational frequencies of CHO and calculated results by density functional (B3LYP and B3PW91) and HF methods indicates that B3LYP is superior to the scaled HF and B3PW91 approach for molecular vibrational problems. Moreover, (13)C NMR and (1)H NMR chemical shifts are calculated by using the gauge independent atomic orbital (GIAO) method with HF/B3LYP/B3PW91 methods and the same basis set. A study on the electronic properties; absorption wavelengths, excitation energy, dipole moment and frontier molecular orbital energies, are performed by HF and DFT methods. The calculated HOMO and LUMO energies show that charge transfer occurs within the molecule. Besides frontier molecular orbitals (FMO), molecular electrostatic potential (MEP) was performed. NLO properties and Mulliken charges of the CHO was also calculated and interpreted. The thermodynamic properties (heat capacity, entropy, and enthalpy) of the title compound at different temperatures are calculated in gas phase.
Sigma-Aldrich
Cyclohexanone oxime, 97%
Pricing and availability is not currently available.