MilliporeSigma
The essential role of PIM kinases in sarcoma growth and bone invasion.
Carcinogenesis (2012-05-25)
Maja Narlik-Grassow, Carmen Blanco-Aparicio, Yolanda Cecilia, Sandra Peregrina, Beatriz Garcia-Serelde, Sandra Muñoz-Galvan, Marta Cañamero, Amancio Carnero
ABSTRACT
PIM kinases are a family of serine/threonine kinases composed of three different isoforms (PIM1, PIM 2 and PIM 3) that are highly homologous. Their expression is mediated by the JAK/STAT signalling pathway, providing survival and cell cycle transition signals. PIM kinases are heavily targeted for anticancer drug discovery. However, very little is known about the relative contribution of the different isoforms to the tumourigenesis process in vivo, and how their individual inhibition might affect tumour growth. Taking advantage of genetically modified mice, we explored whether the inhibition of specific isoforms is required to prevent sarcomas induced by 3-methylcholanthrene carcinogenic treatment. We found that absence of Pim2 and Pim3 greatly reduced sarcoma growth to a similar extent to the absence of all three isoforms. This model of sarcoma generally produces bone invasion by the tumour cells. Lack of Pim2 and Pim3 reduced tumour-induced bone invasion by 70%, which is comparable with the reduction of tumour-induced bone invasion in the absence of all three isoforms. Similar results were obtained in mouse embryonic fibroblasts (MEFs) derived from these knockout (KO) mice, where double Pim2/3 KO MEFs already showed reduced proliferation and were resistant to oncogenic transformation by the RAS oncogene. Our data also suggest an important role of Gsk3β phosphorylation in the process of tumourigenesis.
Product Description
Supelco
3-Methylcholanthrene, analytical standard
Sigma-Aldrich
3-Methylcholanthrene, 98%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Supelco
3-Methylcholanthrene solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid