MilliporeSigma
  • Gender-specific reduction of hepatic Mrp2 expression by high-fat diet protects female mice from ANIT toxicity.
Gender-specific reduction of hepatic Mrp2 expression by high-fat diet protects female mice from ANIT toxicity.
Toxicology and applied pharmacology (2012-04-24)
Bo Kong, Iván L Csanaky, Lauren M Aleksunes, Meghan Patni, Qi Chen, Xiaochao Ma, Hartmut Jaeschke, Scott Weir, Melinda Broward, Curtis D Klaassen, Grace L Guo
ABSTRACT
Emerging evidence suggests that feeding a high-fat diet (HFD) to rodents affects the expression of genes involved in drug transport. However, gender-specific effects of HFD on drug transport are not known. The multidrug resistance-associated protein 2 (Mrp2, Abcc2) is a transporter highly expressed in the hepatocyte canalicular membrane and is important for biliary excretion of glutathione-conjugated chemicals. The current study showed that hepatic Mrp2 expression was reduced by HFD feeding only in female, but not male, C57BL/6J mice. In order to determine whether down-regulation of Mrp2 in female mice altered chemical disposition and toxicity, the biliary excretion and hepatotoxicity of the Mrp2 substrate, α-naphthylisothiocyanate (ANIT), were assessed in male and female mice fed control diet or HFD for 4weeks. ANIT-induced biliary injury is a commonly used model of experimental cholestasis and has been shown to be dependent upon Mrp2-mediated efflux of an ANIT glutathione conjugate that selectively injures biliary epithelial cells. Interestingly, HFD feeding significantly reduced early-phase biliary ANIT excretion in female mice and largely protected against ANIT-induced liver injury. In summary, the current study showed that, at least in mice, HFD feeding can differentially regulate Mrp2 expression and function and depending upon the chemical exposure may enhance or reduce susceptibility to toxicity. Taken together, these data provide a novel interaction between diet and gender in regulating hepatobiliary excretion and susceptibility to injury.
Sigma-Aldrich
1-Naphthyl isothiocyanate, 95%