MilliporeSigma
  • Enaminones 12. An explanation of anticonvulsant activity and toxicity per Linus Pauling's clathrate hypothesis.
Enaminones 12. An explanation of anticonvulsant activity and toxicity per Linus Pauling's clathrate hypothesis.
European journal of medicinal chemistry (2012-03-16)
Patrice L Jackson, Clive D Hanson, Alanna K Farrell, Raymond J Butcher, James P Stables, Natalie D Eddington, K R Scott
ABSTRACT
The x-ray crystal structure of 3-((5-methylisoxazol-3-yl)amino)-5-methylcyclohex-2-enone (12b) and 3-((5-methylisoxazolyl-3-yl)amino)-5,5-dimethylcyclohex-2-enone (12c) were determined and correlated to their anticonvulsant activity in mice and rats. A hypothesis for the toxicity of the analogs are advanced. In addition, a series of 5-methyl-N-(3-oxocyclohex-1-enyl)-isoxazole-3-carboxamides were synthesized and evaluated for anticonvulsant activity. These compounds were compared to the activity of the corresponding amino and aminomethyl enaminones. Additional investigation involved the synthesis and evaluation of a trifluoromethyl analog of the active isoxazole tert-butyl 4-(5-methisoxazol-3-yl-amino)-6-methyl-2-oxo-cyclohex-3-ene carboxylate (4f).