MilliporeSigma
  • Tentative identification of phase I metabolites of HU-210, a classical synthetic cannabinoid, by LC-MS/MS.
Tentative identification of phase I metabolites of HU-210, a classical synthetic cannabinoid, by LC-MS/MS.
Journal of pharmaceutical and biomedical analysis (2012-03-13)
Unyong Kim, Ming Ji Jin, Jaeick Lee, Sang Beom Han, Moon Kyo In, Hye Hyun Yoo
ABSTRACT
(6aR,10aR)-9-(Hydroxymethyl)-6,6-dimethyl-3-(2-methyloctan-2-yl)-6a,7,10,10a-tetrahydrobenzo[c]chromen-1-ol (HU-210) is a synthetic cannabinoid, with a classical cannabinoid structure similar to Δ(9)-tetrahydrocannabinol (Δ(9)-THC). In this study, the in vitro metabolism of HU-210 was investigated in human liver microsomes to characterize associated phase I metabolites. HU-210 was incubated with human liver microsomes, and the reaction mixture was analyzed using LC-MS/MS. HU-210 was metabolized in human liver microsomes, yielding about 24 metabolites. These metabolites were structurally characterized on the basis of accurate mass analyses and MS/MS fragmentation patterns. The major metabolic route for HU-210 was oxygenation. Metabolites M1-M7 were identified as mono-oxygenated metabolites; M8-M15, mono-hydroxylated metabolites; M16-M20, di-oxygenated metabolites; and M21-M24, di-hydroxylated metabolites. These results provide evidence for in vivo HU-210 metabolism, and they may be applied to the analysis of HU-210 and its relevant metabolites in biological samples.
Product Description
Supelco
HU-210 solution, 100 μg/mL in methanol, (Spice Cannabinoid), ampule of 1 mL, certified reference material, Cerilliant®